Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008287.3 | NP_032313.2 | 1 | 125 | |
GenBank mRNA | BC092375.1 | - | 1 | 59 | |
BC125590.1 | - | 1 | 25 | 40130105 | |
BC125592.1 | - | 1 | 25 | 40130107 | |
BU559752.1 | - | 1 | 128 |