Gene Transcripts

Gene Symbol
Hoxd12
Entrez Gene ID
15432
Gene Name
homeobox D12
Gene Aliases
Hox-4.7, Hox-5.6
Chromosome Location
Chr. 2: 74675013 - 74677705 on Build GRCm38
UniGene ID
Mm.57124
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008274.3 NP_032300.2 2 775
GenBank mRNA AK031206.1 - 2 714
BC145656.1 - 2 775 40130959

View Full Product Details