Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008467.4 | NP_032493.1 | 3 | 446 | |
GenBank mRNA | AF020771.1 | - | 3 | 154 | |
AK147920.1 | - | 3 | 455 | ||
BC026821.1 | - | 3 | 241 | 3672708 | |
BC052162.1 | - | 3 | 447 | 6510795 | |
CT010281.1 | - | 3 | 154 |