Gene Transcripts

Gene Symbol
Ltf
Entrez Gene ID
17002
Gene Name
lactotransferrin
Gene Aliases
Csp82, Lf, MMS10R, Ms10r
Chromosome Location
Chr. 9: 111019292 - 111042766 on Build GRCm38
UniGene ID
Mm.282359
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008522.3 NP_032548.2 14 1708
GenBank mRNA AK036491.1 - 14 1709
AK144556.1 - 14 1709
AK150799.1 - 14 1709
AK151822.1 - 14 1709
AK164781.1 - 14 1710
BC006904.1 - 14 1716 3600033
BC008530.1 - 14 1689
BC009662.1 - 10 1139
CT010339.1 - 14 1674
D88510.1 - 14 1708
FJ538998.1 - 15 1674
J03298.1 - 14 1677

View Full Product Details