Gene Transcripts

Gene Symbol
Nab2
Entrez Gene ID
17937
Gene Name
Ngfi-A binding protein 2
Gene Aliases
AI451907
Chromosome Location
Chr. 10: 127660918 - 127666703 on Build GRCm38
UniGene ID
Mm.336898
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122895.1 NP_001116367.1 3, 4 1344
NM_008668.2 NP_032694.2 3, 4 1344
GenBank mRNA AK158688.1 - 3, 4 1274
AK171180.1 - 3, 4 1224
BC045139.1 - 3, 4 1228 6306757
U47543.1 - 3, 4 1319

View Full Product Details