Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008770.3 | NP_032796.1 | 3 | 957 | |
GenBank mRNA | AK005088.1 | - | 3 | 932 | |
AK005171.1 | - | 3 | 930 | ||
AK161698.1 | - | 3 | 932 | ||
BC021659.1 | - | 3 | 909 | 4190031 | |
U19582.1 | - | 3 | 885 |