Gene Transcripts

Gene Symbol
Atxn1
Entrez Gene ID
20238
Gene Name
ataxin 1
Gene Aliases
2900016G23Rik, Atx1, C85907, ENSMUSG00000074917, Gm10786, Sca1
Chromosome Location
Chr. 13: 45549755 - 45965036 on Build GRCm38
UniGene ID
Mm.342683
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199304.1 NP_001186233.1 6 1583
NM_001199305.1 NP_001186234.1 7 1708
NM_009124.6 NP_033150.2 7 1708
GenBank mRNA AK048268.1 - 7 1695
AK169931.1 - 7 1702
BC058178.1 - 6 1549 5326559
X83542.1 - 6 1449

View Full Product Details