Gene Transcripts
- Gene Symbol
- Ssty1
- Entrez Gene ID
- 20611
- Gene Name
- spermiogenesis specific transcript on the Y 1
- Gene Aliases
- Smy, Ssty
- Chromosome Location
- Chr. Y: 18089167 - 18091551 on Build GRCm38
- UniGene ID
- Mm.347498
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009220.2 |
NP_033246.2
|
3 |
696 |
|
GenBank mRNA |
X05260.1 |
- |
3 |
696 |
|
- Gene Symbol
- Gm20831
- Entrez Gene ID
- 100040223
- Gene Name
- predicted gene, 20831
- Gene Aliases
- Chromosome Location
- Chr. Y: 17965160 - 17967601 on Build GRCm38
- UniGene ID
- Mm.387262
- Species
- Mus musculus
View Full Product Details