Gene Transcripts

Gene Symbol
Inmt
Entrez Gene ID
21743
Gene Name
indolethylamine N-methyltransferase
Gene Aliases
Temt
Chromosome Location
Chr. 6: 55170627 - 55174990 on Build GRCm38
UniGene ID
Mm.299
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009349.3 NP_033375.1 2 246
GenBank mRNA AK002281.1 - 2 294
AK013010.1 - 2 283
BC013518.1 - 2 272 4236077
M88694.1 - 2 247

View Full Product Details