Gene Transcripts

Gene Symbol
Tnnc1
Entrez Gene ID
21924
Gene Name
troponin C, cardiac/slow skeletal
Gene Aliases
AI874626, TnC, cTnC, cTnI, tncc
Chromosome Location
Chr. 14: 31208312 - 31211711 on Build GRCm38
UniGene ID
Mm.439921
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009393.2 NP_033419.1 5 465
GenBank mRNA BC061172.1 - 5 459
M29793.1 - 5 465

View Full Product Details