Gene Transcripts

Gene Symbol
Zfp2
Entrez Gene ID
22678
Gene Name
zinc finger protein 2
Gene Aliases
9930007F06Rik, Fnp-2, Zfp-2, mKR2, mkr-2, mszf87
Chromosome Location
Chr. 11: 50898712 - 50916176 on Build GRCm38
UniGene ID
Mm.259230
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044697.2 NP_001038162.1 6 750
NM_001044698.2 NP_001038163.1 6 746
NM_001044700.2 NP_001038165.1 6 702
NM_001294323.1 NP_001281252.1 6 709
NM_178447.3 NP_848542.1 7 787
GenBank mRNA AF483520.1 - 1 150
AF483521.1 - 1 150
AK036776.1 - 7 777
AK131978.1 - 6 697
AK141471.1 - 6 726
BC052029.1 - 4 450
BC057171.1 - 6 716
BC089506.1 - 6 681 30247022
Y00850.1 - 4 582

View Full Product Details