Gene Transcripts
- Gene Symbol
- Aebp1
- Entrez Gene ID
- 11568
- Gene Name
- AE binding protein 1
- Gene Aliases
- ACLP
- Chromosome Location
- Chr. 11: 5861853 - 5872225 on Build GRCm38
- UniGene ID
- Mm.3317
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF053943.1 |
- |
4, 5 |
924 |
|
AK128980.1 |
- |
4, 5 |
1000 |
|
AK158276.1 |
- |
4, 5 |
948 |
|
AK159330.1 |
- |
4, 5 |
928 |
|
AK159342.1 |
- |
4, 5 |
947 |
|
AK159377.1 |
- |
4, 5 |
918 |
|
AK159409.1 |
- |
4, 5 |
948 |
|
AK159774.1 |
- |
4, 5 |
919 |
|
AK159957.1 |
- |
4, 5 |
919 |
|
AK178266.1 |
- |
1, 2 |
71 |
|
AK197857.1 |
- |
1, 2 |
71 |
|
AK207123.1 |
- |
2, 3 |
148 |
|
AK217686.1 |
- |
2, 3 |
148 |
|
BC082577.1 |
- |
4, 5 |
936 |
30648530 |
View Full Product Details