Gene Transcripts

Gene Symbol
Aire
Entrez Gene ID
11634
Gene Name
autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermal dystrophy)
Gene Aliases
Chromosome Location
Chr. 10: 78030022 - 78043610 on Build GRCm38
UniGene ID
Mm.35300
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271549.1 NP_001258478.1 10 1210
NM_001271550.1 NP_001258479.1 10 1201
NM_001271551.1 NP_001258480.1 10 1198
NM_009646.2 NP_033776.1 10 1213
GenBank mRNA AF079536.1 - 10 1183
AF128115.1 - 10 1180
AF128116.1 - 10 1171
AF128117.1 - 10 1168
AF128772.1 - 10 1126
AF128773.1 - 10 1126
AJ132243.1 - 10 1126
AJ243821.1 - 10 1126
AK153690.1 - 11 1264
BC103510.1 - 11 1230 40039055
BC103518.1 - 10 1172 40039058
EU625343.3 - 10 1143
JF267399.1 - 10 1143

View Full Product Details