Gene Transcripts

Gene Symbol
Cdkn1b
Entrez Gene ID
12576
Gene Name
cyclin-dependent kinase inhibitor 1B
Gene Aliases
AA408329, AI843786, Kip1, p27, p27Kip1
Chromosome Location
Chr. 6: 134920401 - 134925525 on Build GRCm38
UniGene ID
Mm.2958
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK046676.1 - 2 878
AK047669.1 - 2 1144
AK050240.1 - 2 1130
AK158411.1 - 2 1123

View Full Product Details