Gene Transcripts

Gene Symbol
CASP10
Entrez Gene ID
843
Gene Name
caspase 10
Gene Aliases
ALPS2, FLICE2, MCH4
Chromosome Location
Chr. 2: 201182881 - 201229406 on Build GRCh38
UniGene ID
Hs.5353
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001206524.1 NP_001193453.1 2 645
NM_001206542.1 NP_001193471.1 2 645
NM_001230.4 NP_001221.2 2 645
NM_001306083.1 NP_001293012.1 2 662
NM_032974.4 NP_116756.2 2 645
NM_032976.3 NP_116758.1 2 645
NM_032977.3 NP_116759.2 2 645
XM_005246907.2 XP_005246964.1 2 431
XM_006712796.3 XP_006712859.1 2 658
XM_011511990.2 XP_011510292.1 2 431
GenBank mRNA AF111344.1 - 1 227
AF111345.1 - 1 227
AJ487678.1 - 2 351
AJ487679.1 - 2 348
AK225475.1 - 2 383
AK304859.1 - 2 402
AK310746.1 - 2 431
AY690601.3 - 2 383
BC042844.1 - 2 381 5176698
KU177959.1 - 1 227
KU177960.1 - 1 227
U60519.1 - 2 374
U86214.1 - 2 421

View Full Product Details