Gene Transcripts

Gene Symbol
OXGR1
Entrez Gene ID
27199
Gene Name
oxoglutarate receptor 1
Gene Aliases
GPR80, GPR99, P2RY15, P2Y15, aKGR
Chromosome Location
Chr. 13: 96985719 - 96995842 on Build GRCh38
UniGene ID
Hs.352218
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080818.3 NP_543008.3 4 400
XM_005254043.3 XP_005254100.1 4 686
XM_005254044.3 XP_005254101.1 4 1707
XM_005254045.2 XP_005254102.1 4 447
XM_005254046.2 XP_005254103.1 4 537
XM_011521077.2 XP_011519379.1 4 397
GenBank mRNA AF370886.1 - 1 174
AJ305372.1 - 1 173
AK122639.1 - 4 400
AK313684.1 - 4 454
BC103881.1 - 1 173 40004608
BC103882.1 - 1 173 40004610
BC103883.1 - 1 173 40004611
JQ308179.1 - 1 156

View Full Product Details