Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010303.3 | NP_034433.3 | 1 | 284 | |
GenBank mRNA | AK011851.1 | - | 1 | 247 | |
AK149766.1 | - | 1 | 178 | ||
BC057665.1 | - | 1 | 138 | 4918717 | |
CJ180381.1 | - | 1 | 284 | ||
M63660.1 | - | 1 | 232 |