Gene Transcripts

Gene Symbol
Gp1ba
Entrez Gene ID
14723
Gene Name
glycoprotein 1b, alpha polypeptide
Gene Aliases
GPIba, GPIbalpha
Chromosome Location
Chr. 11: 70636651 - 70644030 on Build GRCm38
UniGene ID
Mm.377085
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010326.2 NP_034456.2 2 655
GenBank mRNA BC137765.1 - 2 635

View Full Product Details