Gene Transcripts

Gene Symbol
Gzmg
Entrez Gene ID
14944
Gene Name
granzyme G
Gene Aliases
Ctla-7, Ctla7
Chromosome Location
Chr. 14: 56156581 - 56159579 on Build GRCm38
UniGene ID
Mm.14868
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010375.2 NP_034505.1 4 582
GenBank mRNA BC099473.1 - 4 582
J02872.1 - 4 568
M36900.1 - 3 377
X14092.1 - 3 377

View Full Product Details