Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001177883.1 | NP_001171354.1 | 2 | 689 | |
NM_010486.2 | NP_034616.1 | 2 | 284 | ||
NM_207685.1 | NP_997568.1 | 3 | 396 | ||
NM_207686.1 | NP_997569.1 | 2 | 284 | ||
GenBank mRNA | AK141919.1 | - | 2 | 565 | |
AK149201.1 | - | 2 | 501 | ||
AY035378.1 | - | 1 | 133 | ||
AY035379.1 | - | 1 | 133 | ||
AY035380.1 | - | 1 | 133 | ||
BC046598.1 | - | 2 | 208 | 6312245 | |
BC049125.1 | - | 3 | 396 | 6821133 | |
BC058393.1 | - | 3 | 396 | 6821133 | |
U29088.1 | - | 2 | 283 |