Gene Transcripts

Gene Symbol
Pax9
Entrez Gene ID
18511
Gene Name
paired box 9
Gene Aliases
Pax-9
Chromosome Location
Chr. 12: 56690898 - 56712824 on Build GRCm38
UniGene ID
Mm.5035
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC005794.1 - 2 581
BC006930.1 - 1 1263
X84000.1 - 2 613

View Full Product Details