Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011107.1 | NP_035237.1 | 2 | 155 | |
GenBank mRNA | AF094610.1 | - | 2 | 148 | |
AF097637.1 | - | 2 | 136 | ||
AF162712.1 | - | 2 | 155 | ||
AF187852.1 | - | 2 | 134 | ||
AK007730.1 | - | 2 | 149 | ||
AK007797.1 | - | 2 | 154 | ||
AK008331.1 | - | 2 | 151 | ||
AK008664.1 | - | 2 | 151 | ||
AK008668.1 | - | 2 | 151 | ||
AK008841.1 | - | 2 | 149 | ||
AK008934.1 | - | 2 | 155 | ||
AK008936.1 | - | 2 | 149 | ||
AK009314.1 | - | 2 | 148 | ||
AK019009.1 | - | 2 | 149 | ||
AK028104.1 | - | 2 | 152 | ||
AK028134.1 | - | 2 | 155 | ||
BC145908.1 | - | 2 | 155 | 40131211 | |
BC145910.1 | - | 2 | 155 | 40131213 |