Gene Transcripts

Gene Symbol
Thbs4
Entrez Gene ID
21828
Gene Name
thrombospondin 4
Gene Aliases
TSP4
Chromosome Location
Chr. 13: 92751586 - 92794818 on Build GRCm38
UniGene ID
Mm.20865
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011582.3 NP_035712.1 3 568
GenBank mRNA AF102887.1 - 3 508
BC139414.1 - 3 448 8862436

View Full Product Details