Gene Transcripts

Gene Symbol
Clec3b
Entrez Gene ID
21922
Gene Name
C-type lectin domain family 3, member b
Gene Aliases
Tna
Chromosome Location
Chr. 9: 123150946 - 123157432 on Build GRCm38
UniGene ID
Mm.34588
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011606.2 NP_035736.2 2 292
GenBank mRNA BC035043.1 - 2 244
BY114395.1 - 2 292
U08595.1 - 2 275
X79199.1 - 2 267

View Full Product Details