Gene Transcripts

Gene Symbol
Gmcl1
Entrez Gene ID
23885
Gene Name
germ cell-less homolog 1 (Drosophila)
Gene Aliases
2810049L19Rik, Gcl, mglc-1
Chromosome Location
Chr. 6: 86691768 - 86733378 on Build GRCm38
UniGene ID
Mm.321452
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011818.3 NP_035948.3 4 705
GenBank mRNA AF163665.1 - 4 649
AF186095.1 - 4 541
AF282322.1 - 4 575
AK004716.1 - 4 681
AK132788.1 - 4 569
AK146964.1 - 4 674
AK155238.1 - 4 688
BC054767.1 - 4 664 6414943

View Full Product Details