Gene Transcripts

Gene Symbol
Xcr1
Entrez Gene ID
23832
Gene Name
chemokine (C motif) receptor 1
Gene Aliases
Ccxcr1, Gpr5, mXcr1
Chromosome Location
Chr. 9: 123852315 - 123862029 on Build GRCm38
UniGene ID
Mm.390241
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011798.4 NP_035928.2 2 1338
GenBank mRNA AK040878.1 - 2 1339
AK089908.1 - 2 1339
BC141368.1 - 1 1245 9055981
BC141369.1 - 1 1245 9055982

View Full Product Details