Gene Transcripts

Gene Symbol
C2
Entrez Gene ID
12263
Gene Name
complement component 2 (within H-2S)
Gene Aliases
Chromosome Location
Chr. 17: 34862602 - 34882100 on Build GRCm38
UniGene ID
Mm.283217
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013484.2 NP_038512.2 2 292
GenBank mRNA AK146812.1 - 2 293
BC011086.1 - 2 255 3992881
M57891.1 - 2 239

View Full Product Details