Gene Transcripts

Gene Symbol
Tpsg1
Entrez Gene ID
26945
Gene Name
tryptase gamma 1
Gene Aliases
Prss31, Tmt
Chromosome Location
Chr. 17: 25369216 - 25374442 on Build GRCm38
UniGene ID
Mm.180108
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012034.3 NP_036164.2 4 665
GenBank mRNA AF175523.1 - 4 566
AK027982.1 - 3 491
BC019974.1 - 4 589 4215589
BC052325.1 - 4 634 6531021

View Full Product Details