Gene Transcripts

Gene Symbol
Cfd
Entrez Gene ID
11537
Gene Name
complement factor D (adipsin)
Gene Aliases
Adn, DF
Chromosome Location
Chr. 10: 79890853 - 79892660 on Build GRCm38
UniGene ID
Mm.4407
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291915.1 NP_001278844.1 5 884
NM_013459.3 NP_038487.1 5 887
GenBank mRNA AK143817.1 - 5 831
M11768.1 - 5 828

View Full Product Details