Gene Transcripts

Gene Symbol
Psmb10
Entrez Gene ID
19171
Gene Name
proteasome (prosome, macropain) subunit, beta type 10
Gene Aliases
Mecl-1, Mecl1
Chromosome Location
Chr. 8: 105935728 - 105938454 on Build GRCm38
UniGene ID
Mm.787
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013640.3 NP_038668.2 3 603
GenBank mRNA AK144096.1 - 3 326
BC004730.1 - 3 329 3583052
CT010274.1 - 3 221
D85561.1 - 3 298
U77784.1 - 3 290
Y10875.1 - 3 321

View Full Product Details