Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001271730.1 | NP_001258659.1 | 3 | 734 | |
NM_013842.3 | NP_038870.2 | 3 | 734 | ||
GenBank mRNA | AF027963.3 | - | 3 | 408 | |
AF443192.1 | - | 3 | 734 | ||
AK007724.1 | - | 3 | 395 | ||
AK145241.1 | - | 3 | 394 | ||
BC008153.1 | - | 3 | 349 | ||
BC016079.1 | - | 3 | 699 | 4489182 | |
BC029197.1 | - | 3 | 389 | 2648085 | |
BG800705.1 | - | 3 | 365 |