Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018758.2 | NP_061228.1 | 9, 10 | 1769 | |
GenBank mRNA | AF070975.1 | - | 9, 10 | 1723 | |
AK045965.1 | - | 9, 10 | 1770 | ||
AK082046.1 | - | 1689 | |||
AK169462.1 | - | 9, 10 | 1753 | ||
AK170182.1 | - | 9, 10 | 1717 | ||
BC005423.1 | - | 9, 10 | 1701 | 3487448 | |
BC009666.1 | - | 9, 10 | 1713 | 3490710 |