Gene Transcripts

Gene Symbol
Acsl4
Entrez Gene ID
50790
Gene Name
acyl-CoA synthetase long-chain family member 4
Gene Aliases
9430020A05Rik, ACS4, AU018108, Facl4, Lacs4
Chromosome Location
Chr. X: 142317993 - 142390798 on Build GRCm38
UniGene ID
Mm.391337
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033600.1 NP_001028772.1 15 2075
NM_019477.3 NP_062350.3 15 2078
NM_207625.2 NP_997508.1 15 2368
GenBank mRNA AB033885.1 - 15 1887
AB033886.1 - 15 2177
AB033887.1 - 15 2225
AJ243502.1 - 15 1923
AK054387.1 - 15 2048
BC016416.1 - 15 1949 3157957
BC058663.1 - 15 2164 6409748

View Full Product Details