Gene Transcripts
- Gene Symbol
- Pqbp1
- Entrez Gene ID
- 54633
- Gene Name
- polyglutamine binding protein 1
- Gene Aliases
- PQBP-1, Sfc2, npw38
- Chromosome Location
- Chr. X: 7894519 - 7899269 on Build GRCm38
- UniGene ID
- Mm.14616
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001252528.1 |
NP_001239457.1
|
4, 5 |
340 |
|
NM_001252529.1 |
NP_001239458.1
|
4, 5 |
406 |
|
NM_019478.4 |
NP_062351.2
|
4, 5 |
535 |
|
GenBank mRNA |
AJ250406.1 |
- |
3, 4 |
286 |
|
AK077652.1 |
- |
4, 5 |
365 |
|
BC009657.1 |
- |
3, 4 |
308 |
3484619 |
BC051673.1 |
- |
4, 5 |
345 |
30015428 |
BC157976.1 |
- |
4, 5 |
440 |
9088137 |
BQ550428.1 |
- |
4, 5 |
411 |
|
CJ143105.1 |
- |
4, 5 |
341 |
|
CN718083.1 |
- |
3, 4 |
320 |
|
View Full Product Details