Gene Transcripts

Gene Symbol
Arid3b
Entrez Gene ID
56380
Gene Name
AT rich interactive domain 3B (BRIGHT-like)
Gene Aliases
Bdp, Dri2
Chromosome Location
Chr. 9: 57790353 - 57836847 on Build GRCm38
UniGene ID
Mm.425588
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019689.2 NP_062663.1 3 735
GenBank mRNA AF116847.1 - 3 735
AK038068.1 - 3 987
AK084952.1 - 1 47
AK139455.1 - 4 808
AK148073.1 - 4 832
BC049776.1 - 4 957
BC145330.1 - 3 799
BC150755.1 - 3 729 9087666

View Full Product Details