Gene Transcripts

Gene Symbol
Tspan3
Entrez Gene ID
56434
Gene Name
tetraspanin 3
Gene Aliases
1700055K04Rik, TM4-A, Tm4sf8
Chromosome Location
Chr. 9: 56135884 - 56161070 on Build GRCm38
UniGene ID
Mm.28484
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019793.3 NP_062767.3 2 354
GenBank mRNA AB072450.1 - 2 164
AF133427.1 - 2 148
AF242591.1 - 2 347
AK006798.1 - 2 230
AK053325.1 - 2 205
AK089494.1 - 186
AK141951.1 - 2 212
AK146130.1 - 2 342
AK152175.1 - 2 239
AK154919.1 - 186
AK159324.1 - 2 325
AK159418.1 - 2 298
AK159621.1 - 2 309
AK159914.1 - 2 287
AK168231.1 - 2 342
AK169068.1 - 2 325
AK212256.1 - 1 60
AK213393.1 - 1 60
AK213564.1 - 1 60
AK218357.1 - 1 60
BB855376.1 - 2 354
BC043072.1 - 2 332 6404165

View Full Product Details