Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001177373.1 | NP_001170844.1 | 3 | 300 | |
NM_019974.2 | NP_064358.1 | 2 | 266 | ||
GenBank mRNA | AB016226.1 | - | 3 | 150 | |
AB016227.1 | - | 3 | 260 | ||
AK009360.1 | - | 3 | 301 | ||
AK009659.1 | - | 3 | 276 | ||
AK009720.1 | - | 3 | 301 | ||
BC115518.2 | - | 3 | 132 |