Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019921.3 | NP_064305.2 | 8 | 1458 | |
GenBank mRNA | AF021833.1 | - | 5 | 812 | |
AK049399.1 | - | 8 | 1427 | ||
AK051044.1 | - | 6 | 1093 | ||
AK083255.1 | - | 8 | 1427 | ||
AK162966.1 | - | 6 | 1250 | ||
BC038483.1 | - | 5 | 516 | ||
BC054105.1 | - | 5 | 761 | 30022728 |