Gene Transcripts

Gene Symbol
Rec8
Entrez Gene ID
56739
Gene Name
REC8 meiotic recombination protein
Gene Aliases
Rec8L1, mrec
Chromosome Location
Chr. 14: 55618166 - 55625395 on Build GRCm38
UniGene ID
Mm.23149
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020002.3 NP_064386.2 4 489
GenBank mRNA AF262055.1 - 4 419
AK077167.1 - 4 424
AK146832.1 - 4 475
AU080225.1 - 4 490
BC052155.1 - 4 485 6335959

View Full Product Details