Gene Transcripts

Gene Symbol
PKD2L2
Entrez Gene ID
27039
Gene Name
polycystin 2 like 2, transient receptor potential cation channel
Gene Aliases
TRPP5
Chromosome Location
Chr. 5: 137889420 - 137942747 on Build GRCh38
UniGene ID
Hs.716884
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258448.1 NP_001245377.1 2 154
NM_001258449.1 NP_001245378.1 2 154
NM_001300921.1 NP_001287850.1 2 154
NM_014386.3 NP_055201.2 2 154
XM_011543321.1 XP_011541623.1 2 132
XM_017009343.1 XP_016864832.1 2 170
XM_017009344.1 XP_016864833.1 1 322
XM_017009345.1 XP_016864834.1 2 170
GenBank mRNA AF118125.1 - 2 121
AF182034.1 - 2 124
AK301924.1 - 2 129
BC034619.1 - 2 1294
BC038714.1 - 2 134
BC044581.1 - 2 153 4830805
BG719682.1 - 2 120
BG721989.1 - 2 159

View Full Product Details