Gene Transcripts
- Gene Symbol
- Clec2i
- Entrez Gene ID
- 93675
- Gene Name
- C-type lectin domain family 2, member i
- Gene Aliases
- Clr-g, Clrg, Dcl1, LCL-1, OCILrP2
- Chromosome Location
- Chr. 6: 128887573 - 128898167 on Build GRCm38
- UniGene ID
- Mm.391513
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001289706.1 |
NP_001276635.1
|
5, 6 |
545 |
|
NM_001289707.1 |
NP_001276636.1
|
4, 5 |
470 |
|
NM_001289708.1 |
NP_001276637.1
|
3, 4 |
387 |
|
NM_020257.2 |
NP_064653.1
|
4, 5 |
458 |
|
GenBank mRNA |
AF121352.1 |
- |
4, 5 |
431 |
|
AF350411.1 |
- |
4, 5 |
435 |
|
AK041724.1 |
- |
3, 4 |
1173 |
|
AY137344.1 |
- |
4, 5 |
469 |
|
AY137345.1 |
- |
4, 5 |
457 |
|
AY256574.1 |
- |
5, 6 |
536 |
|
AY256575.1 |
- |
4, 5 |
459 |
|
AY256576.1 |
- |
3, 4 |
376 |
|
BC139192.1 |
- |
4, 5 |
431 |
8862214 |
BC139193.1 |
- |
4, 5 |
431 |
8862215 |
BC145574.1 |
- |
4, 5 |
443 |
9054140 |
HQ713440.1 |
- |
3, 4 |
365 |
|
View Full Product Details