Gene Transcripts

Gene Symbol
Sult5a1
Entrez Gene ID
57429
Gene Name
sulfotransferase family 5A, member 1
Gene Aliases
Sult-x1, Sultx1
Chromosome Location
Chr. 8: 123142843 - 123158914 on Build GRCm38
UniGene ID
Mm.6562
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020564.3 NP_065589.2 2 213
GenBank mRNA AF026074.1 - 2 201
AK142243.1 - 2 214
BC012677.1 - 2 209 4187578
CJ065192.1 - 2 214

View Full Product Details