Gene Transcripts

Gene Symbol
Ms4a4b
Entrez Gene ID
60361
Gene Name
membrane-spanning 4-domains, subfamily A, member 4B
Gene Aliases
AI463180, Chandra, Ly116
Chromosome Location
Chr. 19: 11443558 - 11463549 on Build GRCm38
UniGene ID
Mm.33957
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021718.2 NP_068364.1 7 764
GenBank mRNA AF230640.1 - 7 721
AF237913.1 - 7 733
AK039824.1 - 7 739
AK088631.1 - 7 720
AK138737.1 - 7 737
AK172495.1 - 7 737

View Full Product Details