Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021788.2 | NP_068560.1 | 2 | 622 | |
GenBank mRNA | AF075136.1 | - | 2 | 633 | |
AK010928.1 | - | 2 | 623 | ||
AK088745.1 | - | 2 | 627 | ||
BC132081.1 | - | 2 | 605 | 40130358 | |
BC132087.1 | - | 2 | 605 | 40130364 |