Gene Transcripts

Gene Symbol
Pidd1
Entrez Gene ID
57913
Gene Name
p53 induced death domain protein 1
Gene Aliases
1200011D09Rik, AU042446, Lrdd, Pidd
Chromosome Location
Chr. 7: 141438113 - 141444408 on Build GRCm38
UniGene ID
Mm.334321
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022654.1 NP_073145.1 6 1342
GenBank mRNA AF274973.1 - 6 1342
AK004702.1 - 7 1285
AK087880.1 - 1553
AK156863.1 - 8 1330
AK183598.1 - 2 88
BC032202.1 - 6 1303
BC056987.1 - 6 1693
BC089373.1 - 6 1314
BC099686.1 - 6 1680
BC145314.1 - 6 1346
BC145315.1 - 6 1307
BC145857.1 - 6 1307 40131160

View Full Product Details