Gene Transcripts

Gene Symbol
Cacnb2
Entrez Gene ID
12296
Gene Name
calcium channel, voltage-dependent, beta 2 subunit
Gene Aliases
AW060387, CAB2, Cavbeta2, Cchb2
Chromosome Location
Chr. 2: 14604049 - 14988811 on Build GRCm38
UniGene ID
Mm.313930
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023116.4 NP_075605.1 2 301
GenBank mRNA AK034054.1 - 2 302
AK082315.1 - 2 185
BC109156.1 - 2 233 40047524
BC109157.1 - 2 233 40047531
FM872408.1 - 2 300

View Full Product Details