Gene Transcripts
- Gene Symbol
- Eif4e2
- Entrez Gene ID
- 26987
- Gene Name
- eukaryotic translation initiation factor 4E member 2
- Gene Aliases
- 2700069E09Rik, AI036339, AV129531, D0H0S6743E, Eif4el3
- Chromosome Location
- Chr. 1: 87213910 - 87240488 on Build GRCm38
- UniGene ID
- Mm.227183
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039169.1 |
NP_001034258.1
|
5, 6 |
1076 |
|
NM_001039170.1 |
NP_001034259.1
|
5, 6 |
625 |
|
NM_023314.3 |
NP_075803.2
|
5, 6 |
625 |
|
GenBank mRNA |
AF068116.1 |
- |
5, 6 |
539 |
|
AK017221.1 |
- |
5, 6 |
1077 |
|
AK018516.1 |
- |
5, 6 |
623 |
|
AK032965.1 |
- |
5, 6 |
629 |
|
AK076067.1 |
- |
3, 4 |
340 |
|
AK138055.1 |
- |
5, 6 |
564 |
|
AK150495.1 |
- |
5, 6 |
564 |
|
BC028903.1 |
- |
5, 6 |
675 |
4166275 |
BC045153.1 |
- |
5, 6 |
559 |
6441327 |
BC049077.1 |
- |
5, 6 |
593 |
5708556 |
U01137.1 |
- |
5, 6 |
572 |
|
View Full Product Details