Gene Transcripts

Gene Symbol
Zfp84
Entrez Gene ID
74352
Gene Name
zinc finger protein 84
Gene Aliases
2210410P13Rik, 4633401C23Rik, AI415447, BB081605, C86188, KRAB18, Zfp69
Chromosome Location
Chr. 7: 29768552 - 29781419 on Build GRCm38
UniGene ID
Mm.251083
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023750.2 NP_076239.1 5 2763
GenBank mRNA AK034259.1 - 5 2659
AK083748.1 - 5 2704
AK145426.1 - 6 2610

View Full Product Details