Gene Transcripts

Gene Symbol
Ppp6c
Entrez Gene ID
67857
Gene Name
protein phosphatase 6, catalytic subunit
Gene Aliases
2310003C10Rik
Chromosome Location
Chr. 2: 39194369 - 39226338 on Build GRCm38
UniGene ID
Mm.389242
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024209.2 NP_077171.1 1, 2 182
GenBank mRNA AK002764.1 - 1, 2 144
AK009104.1 - 1, 2 162
BC002223.1 - 1, 2 207 3491111

View Full Product Details