Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159730.1 | NP_001153202.1 | 3 | 257 | |
NM_024458.2 | NP_077778.1 | 3 | 245 | ||
GenBank mRNA | AK020798.1 | - | 3 | 246 | |
BC012507.1 | - | 3 | 231 | 4500900 | |
BC021920.1 | - | 3 | 246 | 4506350 | |
BC037176.1 | - | 3 | 257 | ||
BI738229.1 | - | 3 | 248 | ||
L08075.1 | - | 3 | 264 |